Learn More
Cayman Chemical GluR1 PhosphoSer831 Polyclon
SDP
Catalog No. 502886941
Encompass_Preferred
Antigen: phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser831 of GluR1 • Host: rabbit • Cross Reactivity: (+) rat GluR1 • Application(s): WB • The AMPAR are comprised of four distinct GluR subunits designated (GluR1-4), which play key roles in virtually all excitatory neurotransmission in the brain. The GluR1 subunit is potentiated by phosphorylation at Ser831 which has been shown to be mediated by either PKC or CaM kinase II. Phosphorylation of this site has also been linked to synaptic plasticity as well as learning and memory.
Catalog No. 50-288-6941 Supplier Cayman Chemical Supplier No. 106021 EA
Antigen: phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser831 of GluR1 • Host: rabbit • Cross Reactivity: (+) rat GluR1 • Application(s): WB • The AMPAR are comprised of four distinct GluR subunits designated (GluR1-4), which play key roles in virtually all excitatory neurotransmission in the brain. The GluR1 subunit is potentiated by phosphorylation at Ser831 which has been shown to be mediated by either PKC or CaM kinase II. Phosphorylation of this site has also been linked to synaptic plasticity as well as learning and memory.