Learn More
Medchemexpress LLC Glurate (4-acetylbutyric acid) | 3128-06-1 | MFCD00004412 | 130.14 | 10 G
SDP
Catalog No. 5000446010
Encompass_Preferred
Glurate (4-acetylbutyric acid) is an acylating agent used in the development of antiviral compounds. It targets retroviruses such as herpes virus and HIV, and can also be used in the synthesis of acyclic nucleoside derivatives and 5-hydroxyhexanoic acid.
  • Acylating agent
  • Used in development of antiviral compounds
  • Targets retroviruses (herpes virus and HIV)
  • Synthesis of acyclic nucleoside derivatives
  • Synthesis of 5-hydroxyhexanoic acid
Catalog No. 50-004-46010 Supplier Medchemexpress LLC Supplier No. HYW00195710G
Glurate (4-acetylbutyric acid) is an acylating agent used in the development of antiviral compounds. It targets retroviruses such as herpes virus and HIV, and can also be used in the synthesis of acyclic nucleoside derivatives and 5-hydroxyhexanoic acid.
  • Acylating agent
  • Used in development of antiviral compounds
  • Targets retroviruses (herpes virus and HIV)
  • Synthesis of acyclic nucleoside derivatives
  • Synthesis of 5-hydroxyhexanoic acid