Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glut3 protein, human (sf9, His) | >90.0% | 50UG
SDP

Catalog No. 5000174875
Encompass_Preferred

Recombinant full-length human GLUT3 protein produced in Sf9 insect cells and supplied with a C-terminal 10x His tag. The protein is provided as a 0.2 μm filtered solution in HEPES buffer containing a mild detergent, is characterized by a predicted molecular weight of approximately 55.3 kDa and >90.0% purity, and is intended for research use with recommended storage at -80°C.

  • Recombinant full-length human GLUT3 expressed in Sf9 insect cells.
  • C-terminal 10x His tag for affinity purification and detection.
  • Supplied in 20 mM HEPES, pH 7.5, 150 mM NaCl, 0.002% w/v LMNG.
  • Predicted molecular weight ~55.3 kDa and >90.0% purity by reducing SDS-PAGE.
  • Available in multiple microgram quantities for assay flexibility.
  • Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-001-74875 Supplier Medchemexpress LLC Supplier No. HYP70320050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human GLUT3 protein produced in Sf9 insect cells and supplied with a C-terminal 10x His tag. The protein is provided as a 0.2 μm filtered solution in HEPES buffer containing a mild detergent, is characterized by a predicted molecular weight of approximately 55.3 kDa and >90.0% purity, and is intended for research use with recommended storage at -80°C.

  • Recombinant full-length human GLUT3 expressed in Sf9 insect cells.
  • C-terminal 10x His tag for affinity purification and detection.
  • Supplied in 20 mM HEPES, pH 7.5, 150 mM NaCl, 0.002% w/v LMNG.
  • Predicted molecular weight ~55.3 kDa and >90.0% purity by reducing SDS-PAGE.
  • Available in multiple microgram quantities for assay flexibility.
  • Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.