Learn More
Medchemexpress LLC GLUT4-IN-2 | 2454113-83-6 | 99.2% | 385.42 | 25 MG
SDP
Catalog No. 5000362731
Encompass_Preferred
GLUT4-IN-2 is a potent and selective GLUT4 inhibitor with IC50s of 11.4 μM for GLUT1 and 6.8 μM for GLUT4. It induces cell apoptosis and cell cycle arrest at the G0/G1 phase, and demonstrates potent antitumor activity.
  • Potent and selective GLUT4 inhibitor
  • Induces cell apoptosis
  • Induces cell cycle arrest at G0/G1 phase
  • Demonstrates potent antitumor activity
  • Decreases expression of mTOR and CDK2 proteins
  • Increases expression of GRP78 and cleaved caspase 3 proteins
Catalog No. 50-003-62731 Supplier Medchemexpress LLC Supplier No. 25MGHY146980
GLUT4-IN-2 is a potent and selective GLUT4 inhibitor with IC50s of 11.4 μM for GLUT1 and 6.8 μM for GLUT4. It induces cell apoptosis and cell cycle arrest at the G0/G1 phase, and demonstrates potent antitumor activity.
  • Potent and selective GLUT4 inhibitor
  • Induces cell apoptosis
  • Induces cell cycle arrest at G0/G1 phase
  • Demonstrates potent antitumor activity
  • Decreases expression of mTOR and CDK2 proteins
  • Increases expression of GRP78 and cleaved caspase 3 proteins