Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutamate Assay Kit
Catalog No. 502904064
Encompass
Glutamate is an important chemical in general metabolism. It is also a crucial mammalian neurotransmitter that is believed to be involved in a number of neurological and psychiatric disorders such as lateral sclerosis lathyrism autism and Alzheimers disease. Glutamate is also widely used as a flavor enhancer in the food industry. Simple direct and automation-ready procedures for measuring glutamate concentration are very desirable.
Catalog No. 50-290-4064 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3301KT
Glutamate is an important chemical in general metabolism. It is also a crucial mammalian neurotransmitter that is believed to be involved in a number of neurological and psychiatric disorders such as lateral sclerosis lathyrism autism and Alzheimers disease. Glutamate is also widely used as a flavor enhancer in the food industry. Simple direct and automation-ready procedures for measuring glutamate concentration are very desirable.