Learn More
Dojindo Molecular Technologies Inc Glutamate Assay Kit-WST (100 tests) is used to determine intracellular levels of Glutamate with easy to adjust dilutions for standard curve and high stability of stock solution 100 tests
SDP
Catalog No. NC1933284
Encompass_Preferred
Glutamate is not only used for the biosynthesis of proteins and glutathione but also plays an important function as a neurotransmitter. Excess of glutamate is believed to contribute to neurodegenerative diseases such as Alzheimer’s disease. A recent study showed that inhibition of cystine/glutamate transporter (xCT), which takes in cystine and releases glutamate, induces ferroptosis, and current cancer research is exploring xCT as a target. Glutamate Assay Kit-WST enables quantitation of glutamate as a metabolite. This kit has been optimized to quantify glutamate in cell culture supernatants and intracellular glutamate by measuring absorbance in a colorimetric WST reaction. This kit is formatted for 96-well microplates and the assay has a detection limit of 5 μmol/l glutamate. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)
Catalog No. NC1933284 Supplier Dojindo Molecular Technologies Inc Supplier No. G26910
Glutamate is not only used for the biosynthesis of proteins and glutathione but also plays an important function as a neurotransmitter. Excess of glutamate is believed to contribute to neurodegenerative diseases such as Alzheimer’s disease. A recent study showed that inhibition of cystine/glutamate transporter (xCT), which takes in cystine and releases glutamate, induces ferroptosis, and current cancer research is exploring xCT as a target. Glutamate Assay Kit-WST enables quantitation of glutamate as a metabolite. This kit has been optimized to quantify glutamate in cell culture supernatants and intracellular glutamate by measuring absorbance in a colorimetric WST reaction. This kit is formatted for 96-well microplates and the assay has a detection limit of 5 μmol/l glutamate. Dojindo has various quantification kits of Intracellular Metabolites. (https://www.dojindo.com/quantification-of-intracellular-metabolites/)