Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glutamate Dehydrogenase 1 Antibody (YA3026) | 100 UL
SDP

Catalog No. 5000444091
Encompass_Preferred

Glutamate Dehydrogenase 1 antibody (YA3026) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting glutamate dehydrogenase 1.

  • Product type: Antibody
  • Clonality: Recombinant, monoclonal
  • Host species: Rabbit
  • Conjugation: Non-conjugated
  • Clone name: YA3026
  • Reactivity: Human
  • Application summary: Western blot (WB), immunohistochemistry - paraffin section (IHC-P), immunocytochemistry/immunofluorescence (ICC/IF), flow cytometry (FC)

Catalog No. 50-004-44091 Supplier Medchemexpress LLC Supplier No. HYP83281100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutamate Dehydrogenase 1 antibody (YA3026) is a rabbit-derived and non-conjugated IgG monoclonal antibody, targeting glutamate dehydrogenase 1.

  • Product type: Antibody
  • Clonality: Recombinant, monoclonal
  • Host species: Rabbit
  • Conjugation: Non-conjugated
  • Clone name: YA3026
  • Reactivity: Human
  • Application summary: Western blot (WB), immunohistochemistry - paraffin section (IHC-P), immunocytochemistry/immunofluorescence (ICC/IF), flow cytometry (FC)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.