Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glutamate dehydrogenase 1 antibody (YA3026) | 50 UL
SDP

Catalog No. 5000444013
Encompass_Preferred

Glutamate dehydrogenase 1 antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody. It targets Glutamate Dehydrogenase 1 and is suitable for various laboratory applications including Western blot (WB), Immunocytochemistry (ICC/IF), Flow cytometry (FC), and Immunohistochemistry - paraffin section (IHC-P).

  • Derived from domestic rabbit
  • Non-conjugated IgG monoclonal antibody
  • Targets Glutamate Dehydrogenase 1
  • Recombinant clonality
  • Reactivity: human
  • Applications: Western blot (WB), Immunocytochemistry (ICC/IF), Flow cytometry (FC), Immunohistochemistry - paraffin section (IHC-P)

Catalog No. 50-004-44013 Supplier Medchemexpress LLC Supplier No. HYP8328150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutamate dehydrogenase 1 antibody is a rabbit-derived, non-conjugated IgG monoclonal antibody. It targets Glutamate Dehydrogenase 1 and is suitable for various laboratory applications including Western blot (WB), Immunocytochemistry (ICC/IF), Flow cytometry (FC), and Immunohistochemistry - paraffin section (IHC-P).

  • Derived from domestic rabbit
  • Non-conjugated IgG monoclonal antibody
  • Targets Glutamate Dehydrogenase 1
  • Recombinant clonality
  • Reactivity: human
  • Applications: Western blot (WB), Immunocytochemistry (ICC/IF), Flow cytometry (FC), Immunohistochemistry - paraffin section (IHC-P)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.