Learn More
MOLECULAR DEPOT LLC Glutamate Dehydrogenase - 20mg
Catalog No. 5000013331
Encompass_Preferred
Glutamate Dehydrogenase (Bovine Liver) is a high purity (95perc) high specific activity (19U/mg) freeze-dried powder of Glutamate Dehydrogenase (GLDH) a mitochondrial enzyme that catalyzes the oxidative deamination of glutamate into a-ketoglutarate For in vitro research use only
Catalog No. 50-000-13331 Supplier MOLECULAR DEPOT LLC Supplier No. B2010234
Glutamate Dehydrogenase (Bovine Liver) is a high purity (95perc) high specific activity (19U/mg) freeze-dried powder of Glutamate Dehydrogenase (GLDH) a mitochondrial enzyme that catalyzes the oxidative deamination of glutamate into a-ketoglutarate For in vitro research use only