Learn More
TRIBIOSCIENCE INC Glutamate Dehydrogenase (GDH) Colorimetric Assay 100 Tests
SDP
Catalog No. NC2886951
Encompass_Preferred
Tribioscience Glutamate dehydrogenase (GDH) activity Quantification Colorimetric Assay is based on a dehydrogenase coupled reaction convert chromophore to a colored formazan, which can be measured at OD 460 nm. The generated signal is proportional to the content of NADPH. This assay specifically detects GDH activity from the cell lysate. The kit provides the easiest and most accurate approach to measure GDH activity from a variety of samples.
Catalog No. NC2886951 Supplier TRIBIOSCIENCE INC Supplier No. TBS2025
Tribioscience Glutamate dehydrogenase (GDH) activity Quantification Colorimetric Assay is based on a dehydrogenase coupled reaction convert chromophore to a colored formazan, which can be measured at OD 460 nm. The generated signal is proportional to the content of NADPH. This assay specifically detects GDH activity from the cell lysate. The kit provides the easiest and most accurate approach to measure GDH activity from a variety of samples.