Learn More
Sigma Aldrich Fine Chemicals Biosciences Glutamate Dehydrogenase (GlDH), 3000 units
Catalog No. NC0955857
Encompass
Glutamate dehydrogenase has been used to measure residual ammonium by enzymatic analysis during fermentation by wine yeast. It used as a component of cofactor recycling systems for NAD(P) and NAD(P)H.
Catalog No. NC0955857 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10197734001
Glutamate dehydrogenase has been used to measure residual ammonium by enzymatic analysis during fermentation by wine yeast. It used as a component of cofactor recycling systems for NAD(P) and NAD(P)H.