Learn More
Randox Glutamate dehydrogenase (GLDH), 9029-12-3, MFCD00131461, 8x6mL
SDP
Catalog No. NC9737595
Encompass_Preferred
The Randox GLDH (DGKC) assay has a wide measuring range of 2.90 – 77.3 U/l which will comfortably detect levels outside of the healthy range.
Catalog No. NC9737595 Supplier Randox Supplier No. GL441
The Randox GLDH (DGKC) assay has a wide measuring range of 2.90 – 77.3 U/l which will comfortably detect levels outside of the healthy range.