Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamic oxalacetic transaminase, porcine heart, Thermo Scientific Chemicals

Catalog No. AAJ60044JPL
Change view
Click to view available options
Quantity:
1 kU
10 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60044JPL 10 kU
AAJ600444I 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60044JPL Supplier Thermo Scientific Chemicals Supplier No. J60044JPL

Converts L-aspartate and alpha-ketoglutarate to oxaloacetate and L-glutamate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-97-9
MDL Number MFCD00131191
Synonym L-Aspartate: 2-oxoglutarate aminotransferase,Aspartate aminotransferase

Specifications

Packaging Plastic Bottle
Quantity 10 kU
Physical Form Suspension
Chemical Name or Material Glutamic oxalacetic transaminase
EINECSNumber 232-571-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.