Learn More
Cayman Chemical GlutamInase C InhIbItr 11 10mg
SDP
Catalog No. 502942041
Encompass_Preferred
An inhibitor of GAC; inhibits GAC enzyme activity (EC50 = 10.64 nM); reduces the viability of A549 NSCLC cells (IC50 = 4.025 nM); reduces colony formation by, and intracellular glutamate levels in, A549 cells at 0.1 and 1 µM; reduces tumor growth in an A549 mouse xenograft model at 100 mg/kg
Catalog No. 50-294-2041 Supplier Cayman Chemical Supplier No. 3988310 MG
Add to Cart
Add to Cart
An inhibitor of GAC; inhibits GAC enzyme activity (EC50 = 10.64 nM); reduces the viability of A549 NSCLC cells (IC50 = 4.025 nM); reduces colony formation by, and intracellular glutamate levels in, A549 cells at 0.1 and 1 µM; reduces tumor growth in an A549 mouse xenograft model at 100 mg/kg