Learn More
Cayman Chemical Glutaminase C Inhibitor 11, Purity: ≥95%, CAS: 3056437-77-2, 1 mg
SDP
Catalog No. 502942021
Encompass_Preferred
An inhibitor of GAC; inhibits GAC enzyme activity (EC50 = 10.64 nM); reduces the viability of A549 NSCLC cells (IC50 = 4.025 nM); reduces colony formation by, and intracellular glutamate levels in, A549 cells at 0.1 and 1 µM; reduces tumor growth in an A549 mouse xenograft model at 100 mg/kg
Catalog No. 50-294-2021 Supplier Cayman Chemical Supplier No. 398831 MG
An inhibitor of GAC; inhibits GAC enzyme activity (EC50 = 10.64 nM); reduces the viability of A549 NSCLC cells (IC50 = 4.025 nM); reduces colony formation by, and intracellular glutamate levels in, A549 cells at 0.1 and 1 µM; reduces tumor growth in an A549 mouse xenograft model at 100 mg/kg