Learn More
Cayman Chemical GlutamInase-IN-1 10mg
SDP
Catalog No. 502943353
Encompass_Preferred
An inhibitor of GLS1 (IC50 = 1 nM); selective for GLS1 over GDH (IC50 = >13 µM); inhibits the proliferation of A549, H22, Caki-1, and HCT116 cancer cells (IC50s = 0.017, 6.78, 0.019, and 0.009 µM, respectively); induces ROS production in A549 cells at 10 µM; decreases tumor weight in an HCT116 mouse xenograft model at 10 mg/kg
Catalog No. 50-294-3353 Supplier Cayman Chemical Supplier No. 4233910 MG
An inhibitor of GLS1 (IC50 = 1 nM); selective for GLS1 over GDH (IC50 = >13 µM); inhibits the proliferation of A549, H22, Caki-1, and HCT116 cancer cells (IC50s = 0.017, 6.78, 0.019, and 0.009 µM, respectively); induces ROS production in A549 cells at 10 µM; decreases tumor weight in an HCT116 mouse xenograft model at 10 mg/kg