Learn More
Cayman Chemical Glutaminase-IN-3, Purity: ≥98%, CAS: 1439399-45-7, 50 mg
SDP
Catalog No. 502943351
Encompass_Preferred
An inhibitor of GAC (IC50 = 0.24 µM); inhibits the proliferation of PC3 and LNCaP prostate cancer cells (IC50s = 6.14 and 2.13 µM, respectively) and non-cancerous CCD-1072Sk fibroblasts (IC50 = 15.39 µM)
Catalog No. 50-294-3351 Supplier Cayman Chemical Supplier No. 4232550 MG
An inhibitor of GAC (IC50 = 0.24 µM); inhibits the proliferation of PC3 and LNCaP prostate cancer cells (IC50s = 6.14 and 2.13 µM, respectively) and non-cancerous CCD-1072Sk fibroblasts (IC50 = 15.39 µM)