Learn More
Medchemexpress LLC HY-79583 50mg Medchemexpress, Glutaminase-IN-3 CAS:1439399-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7958350MG
Encompass_Preferred
Medchemexpress, HY-79583 50mg Glutaminase-IN-3 CAS:1439399-45-7 Glutaminase-IN-3 (compound 657) is a potent glutaminase inhibitor with an IC50 of 0.24 μM for Glutaminase 1 (GLS1). Glutaminase-IN-3 is extracted from patent WO2014089048A1, compound 657. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2297