Learn More
Cayman Chemical Glutaminase Inhibitor Compound 968, Purity: ≥98%, CAS: 442660-67-5, 1 mg
SDP
Catalog No. 502967730
Encompass_Preferred
An inhibitor of GAC (IC50 = 9.3 µM); has anticancer activity in vitro and in vivo; has antiviral activity in vitro
Catalog No. 50-296-7730 Supplier Cayman Chemical Supplier No. 171991 MG
An inhibitor of GAC (IC50 = 9.3 µM); has anticancer activity in vitro and in vivo; has antiviral activity in vitro