Learn More
Cayman Chemical GlutamInase InhIbItr comp 25mg
SDP
Catalog No. 502965740
Encompass_Preferred
A cell-permeable, reversible inhibitor of a mitochondrial glutaminase splice variant, glutaminase C (IC50 = ~2.5 µM), which is commonly found in cancer cells; blocks transformation induced by Rho GTPases and inhibits the proliferation of cancer cell lines without affecting normal cells
Catalog No. 50-296-5740 Supplier Cayman Chemical Supplier No. 1719925 MG
Add to Cart
Add to Cart
A cell-permeable, reversible inhibitor of a mitochondrial glutaminase splice variant, glutaminase C (IC50 = ~2.5 µM), which is commonly found in cancer cells; blocks transformation induced by Rho GTPases and inhibits the proliferation of cancer cell lines without affecting normal cells