Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glutamine Synthetase Antibody (YA405) | affinity purified | Predicted band size: 42 kDa; Observed band size: 42 kDa | 10 UL
SDP

Catalog No. 5000395783
Encompass_Preferred

Glutamine Synthetase Antibody (YA405) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Glutamine Synthetase. It is supplied in 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% Sodium azide as preservative and appears as a liquid. Glutamine synthetase catalyzes the ATP-dependent conversion of glutamate and ammonia to glutamine, playing a crucial role in ammonia and glutamate detoxification, acid-base homeostasis, cell signaling, and cell proliferation. Glutamine is vital for the biosynthesis of several amino acids, pyrimidines, and purines.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets glutamine synthetase
  • Predicted and observed molecular weight: 42 kDa
  • Affinity purified
  • Stable for 1 year at -20°C (avoid repeated freeze/thaw cycles)
  • Species reactivity: Mouse, Rat
  • Application: Western blot (WB) at 1:500-1:1000 dilution

Catalog No. 50-003-95783 Supplier Medchemexpress LLC Supplier No. HYP8069110UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutamine Synthetase Antibody (YA405) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Glutamine Synthetase. It is supplied in 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% Sodium azide as preservative and appears as a liquid. Glutamine synthetase catalyzes the ATP-dependent conversion of glutamate and ammonia to glutamine, playing a crucial role in ammonia and glutamate detoxification, acid-base homeostasis, cell signaling, and cell proliferation. Glutamine is vital for the biosynthesis of several amino acids, pyrimidines, and purines.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets glutamine synthetase
  • Predicted and observed molecular weight: 42 kDa
  • Affinity purified
  • Stable for 1 year at -20°C (avoid repeated freeze/thaw cycles)
  • Species reactivity: Mouse, Rat
  • Application: Western blot (WB) at 1:500-1:1000 dilution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.