Learn More
ABclonal Technology Glutamine Synthetase GLUL
SDP
Catalog No. 5000005745
Encompass_Preferred
The protein encoded by this gene belongs to the glutamine synthetase family It catalyzes the synthesis of glutamine from glutamate and ammonia in an ATP-dependent reaction This protein plays a role in ammonia and glutamate detoxification acid-base homeostasis cell signaling and cell proliferation Glutamine is an abundant amino acid and is important to the biosynthesis of several amino acids pyrimidines and purines Mutations in this gene are associated with congenital glutamine deficiency and overexpression of this gene was observed in some primary liver cancer samples There are six pseudogenes of this gene found on chromosomes 2 5 9 11 and 12 Alternative splicing results in multiple transcript variants
Catalog No. 50-000-05745 Supplier ABclonal Technology Supplier No. A2185620UL
The protein encoded by this gene belongs to the glutamine synthetase family It catalyzes the synthesis of glutamine from glutamate and ammonia in an ATP-dependent reaction This protein plays a role in ammonia and glutamate detoxification acid-base homeostasis cell signaling and cell proliferation Glutamine is an abundant amino acid and is important to the biosynthesis of several amino acids pyrimidines and purines Mutations in this gene are associated with congenital glutamine deficiency and overexpression of this gene was observed in some primary liver cancer samples There are six pseudogenes of this gene found on chromosomes 2 5 9 11 and 12 Alternative splicing results in multiple transcript variants