Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Glutamine Synthetase Recombinant Protein
SDP

Catalog No. 16594034
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-034 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-034 Supplier ProSci Supplier No. 919700050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

Glutamine Synthetase reglutes intracellular concentration of glutamate. Glutamine Synthetase catalyzes the synthesis of glutamine from glutamate and ammonia. Glutamine is an important source of energy and that takes part in cell prolifetation, inhibition of apoptosis, and cell signaling. Glutamine Synthetase is expressed during early fetal stages, and has a role in maintaining body PH by removing ammonia from circulation. Mutations in the GLUL gene are related to congenital glutamine deficiency.

Specifications

Accession Number P15104
Gene ID (Entrez) 2752
Molecular Weight (g/mol) 43.13 kDa
Quantity 0.05 mg
Source E. coli
Common Name glutamate-ammonia ligase
Gene Symbol GLUL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Sequence Thr2-Asn373
Content And Storage Store at -20°C, stable for 6 months after receipt. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym Glutamine Synthetase, GS, Glutamate Decarboxylase, Glutamate--Ammonia Ligase, GLUL, GLNS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.