Learn More
Medchemexpress LLC HY-112269 5mg Medchemexpress, Glutaminyl Cyclase Inhibitor 1 CAS:2110449-60-8 Purity:>98%
SDP
Catalog No. 501969069
Encompass_Preferred
Medchemexpress, HY-112269 5mg Glutaminyl Cyclase Inhibitor 1 CAS:2110449-60-8 Glutaminyl Cyclase Inhibitor 1 is a glutaminyl cyclase inhibitor with an IC50 of 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9069 Supplier Medchemexpress LLC Supplier No. HY1122695MG
Medchemexpress, HY-112269 5mg Glutaminyl Cyclase Inhibitor 1 CAS:2110449-60-8 Glutaminyl Cyclase Inhibitor 1 is a glutaminyl cyclase inhibitor with an IC50 of 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.