Learn More
TARGETMOL CHEMICALS INC Glutaminyl Cyclase Inhibitor 1
SDP
Catalog No. 502878144
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Glutaminyl Cyclase Inhibitor 1, a glutaminyl cyclase inhibitor with an IC50 of 0.5 uM, can be utilized for studying neurological disorders. Purity 100%
Catalog No. 50-287-8144 Supplier TARGETMOL CHEMICALS INC Supplier No. T114245MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Glutaminyl Cyclase Inhibitor 1, a glutaminyl cyclase inhibitor with an IC50 of 0.5 uM, can be utilized for studying neurological disorders. Purity 100%