Learn More
Cayman Chemical γ-Glutamyl Transferase Activity Assay Kit, 1 ea
SDP
Catalog No. 502892951
Encompass_Preferred
Cayman’s GGT Activity Assay Kit provides a convenient method of detecting GGT activity in plasma, serum, and tissue lysates. In this assay, GGT catalyzes the transfer of the γ-glutamyl group from L-γ-glutamyl-p-nitroaniline (γGpNA) to glycylglycine resulting in the liberation of the colored product p-nitroaniline (pNA), which can be measured at 418 nm. Under circumstances in which the GGT activity rate is limiting, the rate of increase is directly proportional to the GGT activity in the sample.
Catalog No. 50-289-2951 Supplier Cayman Chemical Supplier No. 7022101 EA
Cayman’s GGT Activity Assay Kit provides a convenient method of detecting GGT activity in plasma, serum, and tissue lysates. In this assay, GGT catalyzes the transfer of the γ-glutamyl group from L-γ-glutamyl-p-nitroaniline (γGpNA) to glycylglycine resulting in the liberation of the colored product p-nitroaniline (pNA), which can be measured at 418 nm. Under circumstances in which the GGT activity rate is limiting, the rate of increase is directly proportional to the GGT activity in the sample.