Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-Glutamylphenylalanine | 7432-24-8 | MFCD00002629 | 99.8% | 294.30 | C14H18N2O5 | 25 MG
SDP

Catalog No. 5000213027
Encompass_Preferred

γ-Glu-Phe (γ-Glutamylphenylalanine) analytical standard for research and analytical applications, used as a reference compound in chromatographic assays and flavor chemistry studies. Certificate of analysis is provided with batch-specific identity and purity data.

  • Analytical standard of γ-Glutamylphenylalanine for reference use.
  • Purity by HPLC: 99.83%.
  • Molecular weight: 294.30 g/mol.
  • Chemical formula: C14H18N2O5.
  • CAS number: 7432-24-8.
  • Suitable for chromatographic and flavor analysis applications.
  • Available in small laboratory pack sizes such as 25 MG.

Catalog No. 50-002-13027 Supplier Medchemexpress LLC Supplier No. HY101399R25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ-Glu-Phe (γ-Glutamylphenylalanine) analytical standard for research and analytical applications, used as a reference compound in chromatographic assays and flavor chemistry studies. Certificate of analysis is provided with batch-specific identity and purity data.

  • Analytical standard of γ-Glutamylphenylalanine for reference use.
  • Purity by HPLC: 99.83%.
  • Molecular weight: 294.30 g/mol.
  • Chemical formula: C14H18N2O5.
  • CAS number: 7432-24-8.
  • Suitable for chromatographic and flavor analysis applications.
  • Available in small laboratory pack sizes such as 25 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.