Learn More
Medchemexpress LLC γ-Glutamylphenylalanine | 7432-24-8 | MFCD00002629 | 99.8% | 294.30 g/mol | C14H18N2O5 | 100 MG
SDP
Catalog No. 5000213025
Encompass_Preferred
γ-Glutamylphenylalanine (γ-Glu-Phe) is an analytical reference standard supplied as a high-purity powder for research and quantitative analysis. It has a molecular weight of 294.30 g/mol and CAS 7432-24-8, and is accompanied by batch-specific certificates of analysis and safety data sheets for traceable use in analytical workflows.
  • High purity (≈99.8%) suitable for HPLC and quantitative assays.
  • Certificate of analysis available for batch traceability.
  • CAS-registered compound to aid unambiguous identification.
  • Available in multiple small-package sizes for analytical labs.
  • Provided as a stable powder for long-term storage under recommended conditions.
  • Packaged in amber glass to reduce light exposure.
Catalog No. 50-002-13025 Supplier Medchemexpress LLC Supplier No. HY101399R100MG
γ-Glutamylphenylalanine (γ-Glu-Phe) is an analytical reference standard supplied as a high-purity powder for research and quantitative analysis. It has a molecular weight of 294.30 g/mol and CAS 7432-24-8, and is accompanied by batch-specific certificates of analysis and safety data sheets for traceable use in analytical workflows.
  • High purity (≈99.8%) suitable for HPLC and quantitative assays.
  • Certificate of analysis available for batch traceability.
  • CAS-registered compound to aid unambiguous identification.
  • Available in multiple small-package sizes for analytical labs.
  • Provided as a stable powder for long-term storage under recommended conditions.
  • Packaged in amber glass to reduce light exposure.