Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-Glutamylphenylalanine | 7432-24-8 | MFCD00002629 | 99.8% | 294.30 g/mol | C14H18N2O5 | 100 MG
SDP

Catalog No. 5000213219
Encompass_Preferred

γ-Glu-Phe (γ-glutamylphenylalanine) is an endogenous dipeptide used as a biochemical standard and research reagent. It contains a glutamyl residue linked to phenylalanine via a γ-carboxyl bond and is supplied for studies of peptide metabolism, taste modulation, and analytical method development.

  • High purity: supplied at ~99.8% for reliable experimental results.
  • Well characterized: molecular formula C14H18N2O5 and molecular weight 294.30 g/mol.
  • Multiple pack sizes: available in small to preparative quantities for flexibility.
  • Supported documentation: certificate of analysis and safety data sheet available.
  • Suitable for analytical standards, biochemical assays, and flavor chemistry research.

Catalog No. 50-002-13219 Supplier Medchemexpress LLC Supplier No. HY101399100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ-Glu-Phe (γ-glutamylphenylalanine) is an endogenous dipeptide used as a biochemical standard and research reagent. It contains a glutamyl residue linked to phenylalanine via a γ-carboxyl bond and is supplied for studies of peptide metabolism, taste modulation, and analytical method development.

  • High purity: supplied at ~99.8% for reliable experimental results.
  • Well characterized: molecular formula C14H18N2O5 and molecular weight 294.30 g/mol.
  • Multiple pack sizes: available in small to preparative quantities for flexibility.
  • Supported documentation: certificate of analysis and safety data sheet available.
  • Suitable for analytical standards, biochemical assays, and flavor chemistry research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.