Learn More
Medchemexpress LLC γ-glutamylphenylalanine trifluoroacetate | 2828432-42-2 | 99.5% | 408.33 g/mol | C16H19F3N2O7 | 50 MG

Supplier: Medchemexpress LLC HY101399A50MG
γ-Glu-Phe TFA is the trifluoroacetate salt of the dipeptide gamma-glutamylphenylalanine supplied as a white to off-white solid for research use. It is a high-purity peptide with good aqueous solubility and defined storage recommendations for powdered material and solutions.
- High purity: 99.5%.
- White to off-white solid powder.
- Good aqueous solubility: ~250 mg/mL (may require ultrasonic).
- Powder storage stability: -80°C (2 years) or -20°C (1 year) when sealed and dry.
- In solvent storage stability: -80°C (6 months) or -20°C (1 month) when sealed.
- Suitable for biochemical and flavor research as a gamma-glutamyl dipeptide.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.