Learn More
Electron Microscopy Sciences Glutaraldehyde 2% Paraformaldehyde 2% in Phosphate Buffer, pH 7.4 10 ML
SDP
Catalog No. 50365913
Encompass_Preferred
Glutaraldehyde 2% Paraformaldehyde 2% in Phosphate Buffer.
pH: 7.4
Catalog No. 50-365-913 Supplier Electron Microscopy Sciences Supplier No. 1653606
Glutaraldehyde 2% Paraformaldehyde 2% in Phosphate Buffer.
pH: 7.4