Learn More
Crescent Chemical Co Inc GLUTARALDEHYDE 25%
SDP
Catalog No. 50247656
Encompass_Preferred
Glutaraldehyde 25 % solution in water for electron microscopy, high purity In vials; filled under argon. Size - 10x5ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 23114.02
Catalog No. 50-247-656 Supplier Crescent Chemical Co Inc Supplier No. SE23114.02
Glutaraldehyde 25 % solution in water for electron microscopy, high purity In vials; filled under argon. Size - 10x5ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 23114.02