Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde, 25% Aqueous Solution, MP Biomedicals™

Catalog No. 0219519901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-195-19901 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-195-19901 Supplier MP Biomedicals, Inc Supplier No. 0219519901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutaraldehyde, 25% Aqueous Solution is used in immunohistochemistry (IHC) and tissue processing.

  • Gluteraldehyde is a protein cross-linking agent.
  • It also possesses sporicidal activity.
  • This product is also suitable for use as an electron microscopy fixative.
  • Glutaraldehyde reacts through cross-linking to impart water resistance to protein and polyhydroxy compounds.
  • It is also a reducing agent for photochemicals.
  • In organic syntheses, the reactive carbonyl groups of glutaraldehyde suggest its use as an intermediate for the production of resins, dyestuffs, and pharmaceuticals.
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-30-8
Boiling Point 369 to 372°F at 760 mm Hg (Decomposition)
Melting Point <20°F
Density 1.062 g/mL to 1.124 at 68°F
Synonym Glutaric dialdehyde, 1,3-Diformyl propane, Cidex, Glutarol-1,5-pentanedial, 1,5-Pentanedione, Pentane-1,5-dial
Solubility Information ≥100 mg/mL at 72°F
Molecular Weight (g/mol) 100.117 g/mol
Specific Gravity 1.060 to 1.075
Vapor Pressure 17 mm Hg at 68°F
Vapor Density 3.4
pH 3.1 to 4.5
Quantity 1 mL
Flash Point >95°C (CC)
Physical Form Solution
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.