Learn More
Ted Pella Inc Glutaraldehyde, 25% EM grade, 111-30-8, MFCD00007025, 10 x 10ml
SDP
Supplier:  Ted Pella Inc 18426
Encompass
Molecular Formula C5H8O2, Molecular Weight 100.116, EM grade purified glass distilled Glutaraldehyde. Ampoules are sealed with inert gas.
Catalog No. NC1536477