Learn More
Electron Microscopy Sciences Glutaraldehyde 3% in Sodium Phosphate Buffer, pH 7.2 10 ML
SDP
Catalog No. 50365916
Encompass_Preferred
Glutaraldehyde is available for your convenience in a concentration of 3.0% in Sodium Phosphate Buffer.
pH: 7.2
Catalog No. 50-365-916 Supplier Electron Microscopy Sciences Supplier No. 1653945
Glutaraldehyde is available for your convenience in a concentration of 3.0% in Sodium Phosphate Buffer.
pH: 7.2