Learn More
Crescent Chemical Co Inc GLUTARALDEHYDE 50%
SDP
Supplier:  Crescent Chemical Co Inc SE23116.01
Encompass_Preferred
Glutaraldehyde 50 % solution in water for electron microscopy In vials; filled under argon. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 23116.01
Catalog No. 50-247-659