Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde, 50% aq. soln.

Catalog No. AAA105000E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA105000E 2500 g
AAA1050022 100 g
AAA1050036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA105000E Supplier Thermo Scientific Chemicals Supplier No. A105000E
Only null left
Add to Cart

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaraldehyde is used as a cross linking agent for gelatin, poly(vinyl alcohol) and polyheptapeptides. It is also used as a fixative for electron microscopy and as disinfectants. It provides water resistance to protein and polyhydroxy compounds by reacting through cross linking. It acts as an intermediate for the production of resins, dyestuffs, pharmaceuticals, photographic films and for stabilization of proteins on agarose beads, activation of polystyrene and glass for immobilization of antibodies and antigens and coupling peptides onto carrier proteins.

Solubility
Miscible with water, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

Notes
Incompatible with strong oxidisng agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Melting Point ∼-21°C
Density 1.123
Boiling Point 100°C
Odor Stench
Refractive Index 1.421
Linear Formula OHC(CH2)3CHO
Quantity 2500 g
UN Number UN2922
Beilstein 605390
Merck Index 14,4472
Solubility Information Miscible with water,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene.
Formula Weight 100.12
Concentration or Composition (by Analyte or Components) 50% aq. soln.
Chemical Name or Material Glutaraldehyde
Show More Show Less
Hazard Category H301-H314-H317-H330-H334
Hazard Statement GHS H Statement
H301-H331-H334-H314-H317;
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 203-856-5
RTECSNumber MA2450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.