Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysciences, Inc. Glutaraldehyde Em Gr 25% 100ml. 111-30-8
SDP

Catalog No. NC0445458
Encompass_Preferred

Glutaraldehyde Em Gr 25% 100ml. 111-30-8

Catalog No. NC0445458 Supplier Polysciences, Inc. Supplier No. 01909100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glutaraldehyde Em Gr 25% 100ml. 111-30-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.