Learn More
Med Vet International Glutaraldehyde Cold Sterilization Solution 28 Day 1 Quart
Catalog No. 502832476
Encompass_Preferred
Glutaraldehyde is used as a cold sterilant to disinfect and clean heat-sensitive equipment such as dialysis instruments, surgical instruments, suction bottles, bronchoscopes, endoscopes, and ear, nose, and throat instruments. This chemical can also be used as a tissue fixative in histology and pathology labs and as a hardening agent in the development of x-rays.
Catalog No. 50-283-2476 Supplier Med Vet International Supplier No. MX2805
Glutaraldehyde is used as a cold sterilant to disinfect and clean heat-sensitive equipment such as dialysis instruments, surgical instruments, suction bottles, bronchoscopes, endoscopes, and ear, nose, and throat instruments. This chemical can also be used as a tissue fixative in histology and pathology labs and as a hardening agent in the development of x-rays.