Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde, MP Biomedicals™

Catalog No. ICN19859510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19859510 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19859510 Supplier MP Biomedicals, Inc Supplier No. 0219859510

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-30-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Color Colorless
Density 1.061g/cm3 at 20°C
Quantity 10 mL
Formula Weight 100.11
Physical Form Liquid
Chemical Name or Material Glutaraldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.