Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde Solution (25%/Certified/BioReagent), Fisher BioReagents

Catalog No. BP25481 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25481 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25481

missing translation for 'validMainframeMsg'

Recommended Storage

KEEP REFRIGERATED or in a very cool place to retard polymerisation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-30-8 , 7732-18-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Physical Form Liquid
Melting Point -5°C
Boiling Point 100°C
Density 1.060g/cm³
Packaging Glass Bottle
Vapor Pressure 16.4mmHg at 20°C
Quantity 1 L
Assay Percent Range 24.0 to 26.5 %
Linear Formula HC(O)(CH2)3CHO
Merck Index 15, 4508
Formula Weight 100.12
Percent Purity 24.0 to 26.5%
Grade Certified
Chemical Name or Material Glutaraldehyde Solution, 25%
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Toxic by inhalation. Harmful if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction.
NFPA
Health: 3
Flammability: 1
Instability: 1
DOTInformation DOT Class 6.1, : Poison
Recommended Storage KEEP REFRIGERATED or in a very cool place to retard polymerisation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.