Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde Solution (50%/Certified/BioReagent), Fisher BioReagents

Catalog No. BP25471 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP25471 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25471

missing translation for 'validMainframeMsg'

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

Glass Bottle; 1L

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Chemical Name or Material Glutaraldehyde Solution
Melting Point -33°C
Name Note 50%
Boiling Point 101.5°C
CAS 111-30-8, 7732-18-5
Density 1.130g/cm³
Packaging Glass Bottle
Specific Gravity 1.125 to 1.135 (at 20°C)
Vapor Pressure 15mmHg at 20°C
Viscosity 20 mPa/s (50°C)
Decomposition Information Decomposition Information: 110°C
pH 3.7
Quantity 1 L
Assay Percent Range 49.0 to 52.0 %
Molecular Formula C5H8O2
Linear Formula HC(O)(CH2)3CHO
MDL Number MFCD00007025
UN Number 2927
Merck Index 15, 4508
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
ChEBI CHEBI:64276
Formula Weight 100.12
Percent Purity 49.0 to 52.0%
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.