Learn More
Med Vet International MetriCide 14-Day Sterilizing & Disinfecting Solution, 1 gal.
Catalog No. 502092180
Encompass_Preferred
Glutaraldehyde is used as a cold sterilant to disinfect and clean heat-sensitive equipment such as dialysis instruments, surgical instruments, suction bottles, bronchoscopes, endoscopes, and ear, nose, and throat instruments. This chemical can also be used as a tissue fixative in histology and pathology labs and as a hardening agent in the development of x-rays.
Catalog No. 50-209-2180 Supplier Med Vet International Supplier No. MX1400
Glutaraldehyde is used as a cold sterilant to disinfect and clean heat-sensitive equipment such as dialysis instruments, surgical instruments, suction bottles, bronchoscopes, endoscopes, and ear, nose, and throat instruments. This chemical can also be used as a tissue fixative in histology and pathology labs and as a hardening agent in the development of x-rays.