Learn More
ACROBiosystems Recombinant Protein;Human Glutaredoxin 1 / GLRX1 Protein;E.coli;1MG;GLX,GLRX,GRX,TTase-1,GLRX1,GLRX-1,GRX1,GRX-1
Catalog No. 501658684
Encompass_Preferred
Recombinant Protein;Human Glutaredoxin 1 / GLRX1 Protein;1MG;Human GLRX1, His Tag (GLX-H5149) is expressed from E.coli cells. It contains AA Met 1 - Gln 106 (Accession # NP_001112362).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 12.7 kDa. The protein migrates as 12.7 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8684 Supplier ACROBiosystems Supplier No. GLXH51491MG
Recombinant Protein;Human Glutaredoxin 1 / GLRX1 Protein;1MG;Human GLRX1, His Tag (GLX-H5149) is expressed from E.coli cells. It contains AA Met 1 - Gln 106 (Accession # NP_001112362).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 12.7 kDa. The protein migrates as 12.7 kDa under reducing (R) condition (SDS-PAGE).