Learn More
Medchemexpress LLC Glutaric acid | 110-94-1 | 98.4% | 132.12 | 25 G
SDP
Catalog No. 5000453290
Encompass_Preferred
Glutaric acid is a C5 dicarboxylic acid that serves as an intermediate in the catabolic pathways of lysine and tryptophan. It influences pericyte contractility and migration, and is an indicator for glutaric aciduria type I. In vitro studies demonstrate its ability to reduce TRAP measurement and strongly inhibit GPx activity.
  • Intermediate in catabolic pathways of lysine and tryptophan
  • Influences pericyte contractility and migration
  • Indicator for glutaric aciduria type I
  • Reduces TRAP measurement in a dose-dependent manner
  • Strongly inhibits GPx activity
  • Does not alter Cat and SOD activities
Catalog No. 50-004-53290 Supplier Medchemexpress LLC Supplier No. 25GHYW008820
Glutaric acid is a C5 dicarboxylic acid that serves as an intermediate in the catabolic pathways of lysine and tryptophan. It influences pericyte contractility and migration, and is an indicator for glutaric aciduria type I. In vitro studies demonstrate its ability to reduce TRAP measurement and strongly inhibit GPx activity.
  • Intermediate in catabolic pathways of lysine and tryptophan
  • Influences pericyte contractility and migration
  • Indicator for glutaric aciduria type I
  • Reduces TRAP measurement in a dose-dependent manner
  • Strongly inhibits GPx activity
  • Does not alter Cat and SOD activities