Learn More
Medchemexpress LLC Glutaric acid | 110-94-1 | MFCD00004410 | 98.8% | 132.12 | 25 MG
SDP
Catalog No. 5000451483
Encompass_Preferred
Glutaric acid (Standard) is the analytical standard of Glutaric acid, a C5 dicarboxylic acid that serves as an intermediate during the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I. This product is intended for research and analytical applications. In vitro, Glutaric acid (GA) at concentrations of 1 and 2 mM can reduce TRAP measurement by up to 28% in a dose-dependent manner. It also strongly inhibits the activity of GPx even at lower concentrations, as low as 0.05 mM, in a dose-dependent manner.
  • Intended for research and analytical applications
  • Intermediate in lysine and tryptophan catabolic pathways
  • Affects pericyte contractility and migration
  • Indicator of glutaric aciduria type I
  • Reduces TRAP measurement in a dose-dependent manner
  • Inhibits GPx activity
Catalog No. 50-004-51483 Supplier Medchemexpress LLC Supplier No. 25MGHYW008820R
Glutaric acid (Standard) is the analytical standard of Glutaric acid, a C5 dicarboxylic acid that serves as an intermediate during the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I. This product is intended for research and analytical applications. In vitro, Glutaric acid (GA) at concentrations of 1 and 2 mM can reduce TRAP measurement by up to 28% in a dose-dependent manner. It also strongly inhibits the activity of GPx even at lower concentrations, as low as 0.05 mM, in a dose-dependent manner.
  • Intended for research and analytical applications
  • Intermediate in lysine and tryptophan catabolic pathways
  • Affects pericyte contractility and migration
  • Indicator of glutaric aciduria type I
  • Reduces TRAP measurement in a dose-dependent manner
  • Inhibits GPx activity