Learn More
Medchemexpress LLC Glutaric acid-d4 | 19136-99-3 | MFCD00209727 | 99.46% | 136.14 | 100 MG
SDP
Catalog No. 5000451163
Encompass_Preferred
Glutaric acid-d4 is the deuterium labeled Glutaric acid. Glutaric acid, C5 dicarboxylic acid, is an intermediate during the catabolic pathways of lysine and tryptophan. Glutaric acid affects pericyte contractility and migration. Glutaric acid is an indicator of glutaric aciduria type I.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51163 Supplier Medchemexpress LLC Supplier No. HYW008820S1100MG
Glutaric acid-d4 is the deuterium labeled Glutaric acid. Glutaric acid, C5 dicarboxylic acid, is an intermediate during the catabolic pathways of lysine and tryptophan. Glutaric acid affects pericyte contractility and migration. Glutaric acid is an indicator of glutaric aciduria type I.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.