Learn More
Medchemexpress LLC Glutaric acid-d6 | 154184-99-3 | 98.0% | 138.15 | 25 MG
SDP
Catalog No. 5000366732
Encompass_Preferred
Glutaric acid-d6 is the deuterium labeled Glutaric acid, an intermediate during the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66732 Supplier Medchemexpress LLC Supplier No. 25MGHYW008820S
Add to Cart
Add to Cart
Glutaric acid-d6 is the deuterium labeled Glutaric acid, an intermediate during the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.