Learn More
Medchemexpress LLC Glutaric acid-d6 | 154184-99-3 | MFCD00068286 | 98.0% | 138.15 | 10 MG
SDP
Catalog No. 5000453496
Encompass_Preferred
Glutaric acid-d6 is the deuterium labeled Glutaric acid. Glutaric acid is an intermediate during the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-53496 Supplier Medchemexpress LLC Supplier No. HYW008820S10MG
Glutaric acid-d6 is the deuterium labeled Glutaric acid. Glutaric acid is an intermediate during the catabolic pathways of lysine and tryptophan. It affects pericyte contractility and migration and is an indicator of glutaric aciduria type I.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.