Learn More
Medchemexpress LLC Glutaric acid (Standard) | 110-94-1 | 98.8% | 132.12 | 50 MG
SDP
Catalog No. 5000452131
Encompass_Preferred
Glutaric acid (Standard) is the analytical standard of Glutaric acid, intended for research and analytical applications. This C5 dicarboxylic acid acts as an intermediate in the catabolic pathways of lysine and tryptophan, influences pericyte contractility and migration, and serves as an indicator of glutaric aciduria type I.
  • Reduces TRAP measurement by up to 28% in a dose-dependent manner.
  • Strongly inhibits GPx activity by up to 46% even at low concentrations.
  • Does not alter Cat and SOD activities.
  • Applicable for qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS applications.
  • Features a solid, white to off-white appearance.
  • Chemical formula is C5H8O4.
Catalog No. 50-004-52131 Supplier Medchemexpress LLC Supplier No. HYW008820R50MG
Glutaric acid (Standard) is the analytical standard of Glutaric acid, intended for research and analytical applications. This C5 dicarboxylic acid acts as an intermediate in the catabolic pathways of lysine and tryptophan, influences pericyte contractility and migration, and serves as an indicator of glutaric aciduria type I.
  • Reduces TRAP measurement by up to 28% in a dose-dependent manner.
  • Strongly inhibits GPx activity by up to 46% even at low concentrations.
  • Does not alter Cat and SOD activities.
  • Applicable for qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS applications.
  • Features a solid, white to off-white appearance.
  • Chemical formula is C5H8O4.